Assay Details
| Gene Symbol: Marco | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010766.2 | NP_034896.1 | 9 - 10 | 1165 | 65 | |
| GenBank mRNA | AK156388.1 | - | 9 - 10 | 839 | 65 | |
| AK159967.1 | - | 9 - 10 | 847 | 65 | ||
| AK169230.1 | - | 9 - 10 | 841 | 65 | ||
| BC132338.1 | - | 9 - 10 | 1105 | 40130615 | 65 | |
| BC137939.1 | - | 9 - 10 | 1105 | 8860958 | 65 | |
| U18424.1 | - | 9 - 10 | 1017 | 65 | ||
More Information
Set Membership: |
|

Marco
Genomic Map