Assay Details
| Gene Symbol: Mmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010809.1 | NP_034939.1 | 4 - 5 | 710 | 66 | |
| GenBank mRNA | BC006725.1 | - | 4 - 5 | 674 | 3962288 | 66 |
| X63162.1 | - | 4 - 5 | 710 | 66 | ||
| X66402.1 | - | 4 - 5 | 652 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Complement factor 5 is a quantitative trait gene that modifies liver fibrogenesis in mice and humans.
- Authors
- Hillebrandt S, Wasmuth HE, Weiskirchen R, Hellerbrand C, Keppeler H, Werth A, Schirin-Sokhan R, Wilkens G, Geier A, Lorenzen J, Kohl J, Gressner AM, Matern S, Lammert F
- Journal
- Nat Genet - Page No 835 (Mon Aug 01 00:00:00 EDT 2005)
- Volume (Year)
- 37 (2005)
- Product Usage
- -

Mmp3
Genomic Map