Assay Details
| Gene Symbol: Mmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010809.1 | NP_034939.1 | 5 - 6 | 869 | 90 | |
| GenBank mRNA | AK148467.1 | - | 5 - 6 | 847 | 90 | |
| AY622967.1 | - | 1 - 2 | 74 | 90 | ||
| BC006725.1 | - | 5 - 6 | 833 | 3962288 | 90 | |
| X63162.1 | - | 5 - 6 | 869 | 90 | ||
| X66402.1 | - | 5 - 6 | 811 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp3
Genomic Map