Assay Details
| Gene Symbol: Myd88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010851.2 | NP_034981.1 | 2 - 3 | 547 | 71 | |
| GenBank mRNA | AK150083.1 | - | 2 - 3 | 430 | 71 | |
| AK151748.1 | - | 2 - 3 | 503 | 71 | ||
| AK152596.1 | - | 2 - 3 | 532 | 71 | ||
| AK152935.1 | - | 2 - 3 | 505 | 71 | ||
| AK167929.1 | - | 2 - 3 | 527 | 71 | ||
| AK186240.1 | - | 2 - 3 | 315 | 71 | ||
| BC005591.1 | - | 2 - 3 | 547 | 3482246 | 71 | |
| BC051075.1 | - | 2 - 3 | 494 | 71 | ||
| BC058787.1 | - | 2 - 3 | 482 | 6309465 | 71 | |
| DQ790885.1 | - | 2 - 3 | 466 | 71 | ||
| DQ790886.1 | - | 2 - 3 | 466 | 71 | ||
| U84409.1 | - | 2 - 3 | 528 | 71 | ||
| X51397.1 | - | 2 - 3 | 416 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myd88
Genomic Map