Assay Details
| Gene Symbol: Nek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010892.3 | NP_035022.2 | 2 - 3 | 483 | 78 | |
| GenBank mRNA | AF007247.1 | - | 2 - 3 | 397 | 78 | |
| AF013166.1 | - | 2 - 3 | 367 | 78 | ||
| AK077627.1 | - | 2 - 3 | 447 | 78 | ||
| AK147072.1 | - | 2 - 3 | 459 | 78 | ||
| AK164467.1 | - | 2 - 3 | 426 | 78 | ||
| AK169330.1 | - | 3 - 4 | 374 | 78 | ||
| AK170179.1 | - | 2 - 3 | 403 | 78 | ||
| AK171277.1 | - | 2 - 3 | 403 | 78 | ||
| AK171387.1 | - | 2 - 3 | 403 | 78 | ||
| BC010302.1 | - | 2 - 3 | 405 | 3597227 | 78 | |
| BC011316.1 | - | 2 - 3 | 403 | 3674001 | 78 | |
| BC057576.1 | - | 2 - 3 | 423 | 6810914 | 78 | |
| CF738658.1 | - | 2 - 3 | 510 | 78 | ||
| U95610.1 | - | 2 - 3 | 394 | 78 | ||
More Information
Set Membership: |
|

Nek2
Genomic Map