Assay Details
| Gene Symbol: Nos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010927.3 | NP_035057.1 | 6 - 7 | 794 | 100 | |
| GenBank mRNA | AF065920.1 | - | 6 - 7 | 791 | 100 | |
| AF065921.2 | - | 6 - 7 | 791 | 100 | ||
| AY090567.1 | - | 5 - 6 | 614 | 100 | ||
| BC062378.1 | - | 6 - 7 | 803 | 4978648 | 100 | |
| M84373.1 | - | 6 - 7 | 795 | 100 | ||
| M87039.1 | - | 6 - 7 | 866 | 100 | ||
| U43428.1 | - | 6 - 7 | 682 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nos2
Genomic Map