Assay Details
| Gene Symbol: Nppc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010933.5 | NP_035063.1 | 1 - 2 | 223 | 123 | |
| GenBank mRNA | AK020093.1 | - | 1 - 2 | 226 | 123 | |
| AK036176.1 | - | 1 - 2 | 225 | 123 | ||
| BC111834.1 | - | 1 - 2 | 154 | 40055241 | 123 | |
| BY718381.1 | - | 1 - 2 | 224 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nppc
Genomic Map