Assay Details
| Gene Symbol: Gpr143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010951.3 | NP_035081.3 | 1 - 2 | 375 | 68 | |
| GenBank mRNA | AK033060.1 | - | 1 - 2 | 279 | 68 | |
| BC119384.1 | - | 1 - 2 | 291 | 8734133 | 68 | |
| BC137813.1 | - | 1 - 2 | 291 | 8860832 | 68 | |
| CJ131873.1 | - | 1 - 2 | 376 | 68 | ||
| U63918.1 | - | 1 - 2 | 310 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr143
Genomic Map