Assay Details
| Gene Symbol: Cdk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011049.4 | NP_035179.1 | 2 - 3 | 520 | 114 | |
| GenBank mRNA | AK051425.1 | - | 2 - 3 | 521 | 114 | |
| AK143518.1 | - | 2 - 3 | 548 | 114 | ||
| AK145483.1 | - | 2 - 3 | 550 | 114 | ||
| BC011069.1 | - | 2 - 3 | 263 | 4006745 | 114 | |
| BC013663.1 | - | 2 - 3 | 579 | 4208974 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk16
Genomic Map