Assay Details
| Gene Symbol: Cdk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011049.4 | NP_035179.1 | 4 - 5 | 786 | 89 | |
| GenBank mRNA | AK051425.1 | - | 4 - 5 | 787 | 89 | |
| AK143518.1 | - | 4 - 5 | 814 | 89 | ||
| AK145483.1 | - | 4 - 5 | 816 | 89 | ||
| AK166188.1 | - | 4 - 5 | 995 | 89 | ||
| BC011069.1 | - | 4 - 5 | 529 | 4006745 | 89 | |
| BC013663.1 | - | 4 - 5 | 845 | 4208974 | 89 | |
| X69025.1 | - | 4 - 5 | 808 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk16
Genomic Map