Assay Details
| Gene Symbol: Cdk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011049.4 | NP_035179.1 | 7 - 8 | 1048 | 78 | |
| GenBank mRNA | AK051425.1 | - | 7 - 8 | 1049 | 78 | |
| AK141083.1 | - | 1 - 2 | 67 | 78 | ||
| AK143518.1 | - | 7 - 8 | 1076 | 78 | ||
| AK145483.1 | - | 7 - 8 | 1078 | 78 | ||
| AK166188.1 | - | 7 - 8 | 1257 | 78 | ||
| AK180401.1 | - | 1 - 2 | 101 | 78 | ||
| AK180999.1 | - | 1 - 2 | 101 | 78 | ||
| AK194254.1 | - | 1 - 2 | 101 | 78 | ||
| AK194908.1 | - | 1 - 2 | 101 | 78 | ||
| AK203854.1 | - | 1 - 2 | 101 | 78 | ||
| AK214881.1 | - | 2 - 3 | 164 | 78 | ||
| BC011069.1 | - | 7 - 8 | 791 | 4006745 | 78 | |
| BC013663.1 | - | 7 - 8 | 1107 | 4208974 | 78 | |
| X69025.1 | - | 7 - 8 | 1070 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk16
Genomic Map