Assay Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080773.2 | NP_001074242.1 | 1 - 2 | 155 | 79 | |
| NM_011062.4 | NP_035192.2 | 1 - 2 | 155 | 79 | ||
| GenBank mRNA | AF079535.1 | - | 1 - 2 | 112 | 79 | |
| AF086625.1 | - | 1 - 2 | 121 | 79 | ||
| AF126294.1 | - | 1 - 2 | 96 | 79 | ||
| AK143283.1 | - | 1 - 2 | 118 | 79 | ||
| AK147149.1 | - | 1 - 2 | 147 | 79 | ||
| AK147835.1 | - | 1 - 2 | 130 | 79 | ||
| AK149162.1 | - | 1 - 2 | 88 | 79 | ||
| AK149289.1 | - | 1 - 2 | 113 | 79 | ||
| AK162677.1 | - | 1 - 2 | 115 | 79 | ||
| AK165564.1 | - | 1 - 2 | 92 | 79 | ||
| BC146001.1 | - | 1 - 2 | 92 | 40131304 | 79 | |
| BC146003.1 | - | 1 - 2 | 92 | 40131306 | 79 | |
| BY257456.1 | - | 1 - 2 | 156 | 79 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF079535.1,AF086625.1,AF126294.1,AK147149.1,AK147835.1,AK149289.1,AK162677.1,BC146001.1,BC146003.1,NM_001080773.2,NM_011062.4 transcripts.
Set Membership: |
|

Pdpk1
Genomic Map