Assay Details
| Gene Symbol: Abcb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011075.2 | NP_035205.1 | 16 - 17 | 2205 | 72 | |
| GenBank mRNA | AK030328.1 | - | 15 - 16 | 2148 | 72 | |
| BC141363.1 | - | 16 - 17 | 2162 | 9055975 | 72 | |
| BC150811.1 | - | 16 - 17 | 2162 | 9087722 | 72 | |
| M14757.1 | - | 16 - 17 | 2165 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb1b
Genomic Map