Assay Details
| Gene Symbol: Prkacb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164198.1 | NP_001157670.1 | 6 - 7 | 808 | 72 | |
| NM_001164199.1 | NP_001157671.1 | 6 - 7 | 681 | 72 | ||
| NM_001164200.1 | NP_001157672.1 | 6 - 7 | 773 | 72 | ||
| NM_011100.4 | NP_035230.1 | 6 - 7 | 812 | 72 | ||
| GenBank mRNA | AK048319.1 | - | 6 - 7 | 804 | 72 | |
| AK139419.1 | - | 6 - 7 | 553 | 72 | ||
| AK148728.1 | - | 6 - 7 | 622 | 72 | ||
| AK150122.1 | - | 6 - 7 | 774 | 72 | ||
| AK150134.1 | - | 6 - 7 | 774 | 72 | ||
| AK163585.1 | - | 6 - 7 | 586 | 72 | ||
| AK196882.1 | - | 1 - 2 | 41 | 72 | ||
| BC054533.1 | - | 6 - 7 | 796 | 5704796 | 72 | |
| J02626.1 | - | 6 - 7 | 741 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkacb
Genomic Map