Assay Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011101.3 | NP_035231.2 | 8 - 9 | 1139 | 82 | |
| GenBank mRNA | AK147084.1 | - | 8 - 9 | 1140 | 82 | |
| BC096493.1 | - | 8 - 9 | 933 | 30742571 | 82 | |
| M25811.1 | - | 8 - 9 | 1173 | 82 | ||
| X52684.1 | - | 9 - 10 | 1080 | 82 | ||
| X52685.1 | - | 9 - 10 | 1080 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkca
Genomic Map