Assay Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011101.3 | NP_035231.2 | 10 - 11 | 1450 | 59 | |
| GenBank mRNA | AK147084.1 | - | 10 - 11 | 1451 | 59 | |
| BC096493.1 | - | 10 - 11 | 1244 | 30742571 | 59 | |
| M25811.1 | - | 10 - 11 | 1484 | 59 | ||
| X52684.1 | - | 11 - 12 | 1391 | 59 | ||
| X52685.1 | - | 11 - 12 | 1391 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkca
Genomic Map