Assay Details
| Gene Symbol: Prkcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011103.3 | NP_035233.1 | 3 - 4 | 551 | 79 | |
| GenBank mRNA | AB011812.1 | - | 2 - 3 | 314 | 79 | |
| AF251036.1 | - | 2 - 3 | 314 | 79 | ||
| AK144231.1 | - | 3 - 4 | 556 | 79 | ||
| AK168768.1 | - | 3 - 4 | 546 | 79 | ||
| AY545076.1 | - | 2 - 3 | 314 | 79 | ||
| BC051416.1 | - | 3 - 4 | 515 | 6515302 | 79 | |
| M69042.1 | - | 3 - 4 | 546 | 79 | ||
| X60304.1 | - | 2 - 3 | 327 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkcd
Genomic Map