Assay Details
| Gene Symbol: Prkcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011103.3 | NP_035233.1 | 12 - 13 | 1489 | 70 | |
| GenBank mRNA | AB011812.1 | - | 11 - 12 | 1330 | 70 | |
| AB201453.1 | - | 9 - 10 | 1091 | 70 | ||
| AB201454.1 | - | 9 - 10 | 1169 | 70 | ||
| AB201455.1 | - | 9 - 10 | 1091 | 70 | ||
| AB201456.1 | - | 9 - 10 | 1169 | 70 | ||
| AF251036.1 | - | 11 - 12 | 1252 | 70 | ||
| AK144231.1 | - | 12 - 13 | 1494 | 70 | ||
| AK168768.1 | - | 12 - 13 | 1484 | 70 | ||
| AK171476.1 | - | 12 - 13 | 1492 | 70 | ||
| AY545076.1 | - | 11 - 12 | 1252 | 70 | ||
| BC051416.1 | - | 12 - 13 | 1453 | 6515302 | 70 | |
| M69042.1 | - | 12 - 13 | 1484 | 70 | ||
| X60304.1 | - | 11 - 12 | 1265 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkcd
Genomic Map