Assay Details
| Gene Symbol: Prkcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011103.3 | NP_035233.1 | 13 - 14 | 1586 | 62 | |
| GenBank mRNA | AB011812.1 | - | 12 - 13 | 1427 | 62 | |
| AB201453.1 | - | 10 - 11 | 1188 | 62 | ||
| AB201454.1 | - | 10 - 11 | 1266 | 62 | ||
| AB201455.1 | - | 10 - 11 | 1188 | 62 | ||
| AB201456.1 | - | 10 - 11 | 1266 | 62 | ||
| AF251036.1 | - | 12 - 13 | 1349 | 62 | ||
| AK144231.1 | - | 13 - 14 | 1591 | 62 | ||
| AK168768.1 | - | 13 - 14 | 1581 | 62 | ||
| AK171476.1 | - | 13 - 14 | 1589 | 62 | ||
| AY545076.1 | - | 12 - 13 | 1349 | 62 | ||
| BC051416.1 | - | 13 - 14 | 1550 | 6515302 | 62 | |
| HQ840432.1 | - | 5 - 6 | 440 | 62 | ||
| M69042.1 | - | 13 - 14 | 1581 | 62 | ||
| X60304.1 | - | 12 - 13 | 1362 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkcd
Genomic Map