Assay Details
| Gene Symbol: Plaur | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011113.3 | NP_035243.1 | 1 - 2 | 68 | 72 | |
| GenBank mRNA | AK002580.1 | - | 1 - 2 | 139 | 72 | |
| AK131951.1 | - | 1 - 2 | 84 | 72 | ||
| AK144796.1 | - | 1 - 2 | 85 | 72 | ||
| AK146827.1 | - | 1 - 2 | 83 | 72 | ||
| AK154141.1 | - | 1 - 2 | 85 | 72 | ||
| AK155351.1 | - | 1 - 2 | 72 | 72 | ||
| BC010309.1 | - | 1 - 2 | 68 | 72 | ||
| X62700.1 | - | 1 - 2 | 69 | 72 | ||
| X62701.1 | - | 1 - 2 | 69 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plaur
Genomic Map