| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500006435.mm.1 |
NM_001113418.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006435.mm.1 |
NM_001113418.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006435.mm.1 |
TC1500000821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500006444.mm.1 |
NM_001113418.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006444.mm.1 |
NM_001113418.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006444.mm.1 |
NM_011144.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006444.mm.1 |
NM_011144.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006444.mm.1 |
TC1500000821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500006443.mm.1 |
NM_001113418.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006443.mm.1 |
NM_001113418.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006443.mm.1 |
NM_011144.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006443.mm.1 |
NM_011144.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500006443.mm.1 |
TC1500000821.mm.1 |
MTA TC |