Assay Details
| Gene Symbol: Ppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113418.1 | NP_001106889.1 | 4 - 5 | 572 | 68 | |
| NM_011144.6 | NP_035274.2 | 4 - 5 | 810 | 68 | ||
| GenBank mRNA | AK035676.1 | - | 4 - 5 | 811 | 68 | |
| AK081709.1 | - | 4 - 5 | 573 | 68 | ||
| AK149460.1 | - | 4 - 5 | 509 | 68 | ||
| BC016892.1 | - | 4 - 5 | 803 | 4222384 | 68 | |
| X57638.1 | - | 4 - 5 | 531 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppara
Genomic Map