Assay Details
| Gene Symbol: Ppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113418.1 | NP_001106889.1 | 5 - 6 | 712 | 115 | |
| NM_011144.6 | NP_035274.2 | 5 - 6 | 950 | 115 | ||
| GenBank mRNA | AK035676.1 | - | 5 - 6 | 951 | 115 | |
| AK081709.1 | - | 5 - 6 | 713 | 115 | ||
| AK149460.1 | - | 5 - 6 | 649 | 115 | ||
| BC016892.1 | - | 5 - 6 | 943 | 4222384 | 115 | |
| X57638.1 | - | 5 - 6 | 671 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppara
Genomic Map