Assay Details
| Gene Symbol: Mapk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011161.5 | NP_035291.4 | 2 - 3 | 338 | 60 | |
| GenBank mRNA | AF135185.1 | - | 2 - 3 | 318 | 60 | |
| AK144846.1 | - | 2 - 3 | 311 | 60 | ||
| AK161190.1 | - | 2 - 3 | 226 | 60 | ||
| AK161562.1 | - | 2 - 3 | 290 | 60 | ||
| BC057211.1 | - | 2 - 3 | 274 | 60 | ||
| BC064737.1 | - | 2 - 3 | 289 | 30112690 | 60 | |
| BC092526.1 | - | 2 - 3 | 338 | 6593321 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk11
Genomic Map