Assay Details
| Gene Symbol: Procr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011171.2 | NP_035301.2 | 2 - 3 | 510 | 53 | |
| GenBank mRNA | AK136541.1 | - | 2 - 3 | 370 | 53 | |
| AK143719.1 | - | 2 - 3 | 612 | 53 | ||
| AK167302.1 | - | 2 - 3 | 450 | 53 | ||
| BC028755.1 | - | 2 - 3 | 383 | 1054063 | 53 | |
| CT010175.1 | - | 2 - 3 | 329 | 53 | ||
| L39017.1 | - | 2 - 3 | 384 | 53 | ||
| X63748.2 | - | 2 - 3 | 416 | 53 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Procr
Genomic Map