Assay Details
| Gene Symbol: Procr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011171.2 | NP_035301.2 | 3 - 4 | 796 | 67 | |
| GenBank mRNA | AK136541.1 | - | 3 - 4 | 657 | 67 | |
| AK143719.1 | - | 3 - 4 | 898 | 67 | ||
| AK167302.1 | - | 3 - 4 | 736 | 67 | ||
| CT010175.1 | - | 3 - 4 | 615 | 67 | ||
| L39017.1 | - | 3 - 4 | 670 | 67 | ||
| X63748.2 | - | 3 - 4 | 702 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382152 | X63748.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382152 | AK167302.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382152 | BC028755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382152 | BY393288.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382152 | L39017.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382152 | X63748.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382152 | AK167302.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382152 | BC028755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382152 | BY393288.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382152 | L39017.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970662 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129396 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382152 | X63748.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382152 | AK167302.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382152 | BC028755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382152 | BY393288.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382152 | L39017.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382152 | X63748.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382152 | AK167302.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382152 | BC028755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382152 | BY393288.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382152 | L39017.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382152 | X63748.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382152 | AK167302.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382152 | BC028755.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382152 | BY393288.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382152 | L39017.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382152 | X63748.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382152 | AK167302.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382152 | BC028755.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382152 | BY393288.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382152 | L39017.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21767383 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129396 | - | - |
| Whole Mouse Genome G4122A | A_51_P382152 | X63748.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P382152 | AK167302.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P382152 | BC028755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P382152 | BY393288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P382152 | L39017.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P382152 | X63748.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P382152 | AK167302.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P382152 | BC028755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P382152 | BY393288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P382152 | L39017.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132369 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129396 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02033018 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02128545 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01959859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21767397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02191090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02191094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02191102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01594918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02128540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02017631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01800258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02191104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02191109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01901135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01594913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02191110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01489389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02191086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01489391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01733918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01203364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02191105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01489392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01148215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01778677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21767460 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10477720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477718 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477719 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420664_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10477717 | - | - |
| Mouse Genome 430 2.0 Array | 1420664_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1420664_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19124_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19124_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa14580_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200018981.mm.1 | NM_011171.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018981.mm.1 | NM_011171.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018981.mm.1 | TC0200002374.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420664_PM_s_at | - | - |
| Mouse Expression Set 430 | 1420664_s_at | - | - |
| Murine Genome U74v2 Set | 98018_at | - | - |
| Murine Genome U74v2 Set | 98018_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420664_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420664_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420664_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420664_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4920687 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920687 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2771774 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2771774 | - | - |
| mouse-6 v1.1 (Illumina) | scl20048.3_40-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920687 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20048.3_40-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920687 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2904658 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2771774 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2904658 | - | - |

Procr
Genomic Map