Assay Details
| Gene Symbol: Rab18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181070.6 | NP_851415.1 | 3 - 4 | 287 | 125 | |
| GenBank mRNA | AB232612.1 | - | 3 - 4 | 189 | 125 | |
| AK045514.1 | - | 3 - 4 | 250 | 125 | ||
| AK140901.1 | - | 3 - 4 | 255 | 125 | ||
| AK143673.1 | - | 3 - 4 | 263 | 125 | ||
| AK146177.1 | - | 3 - 4 | 212 | 125 | ||
| AK146397.1 | - | 3 - 4 | 212 | 125 | ||
| AK154920.1 | - | 3 - 4 | 264 | 125 | ||
| BC056351.1 | - | 3 - 4 | 241 | 6807284 | 125 | |
| L04966.1 | - | 3 - 4 | 193 | 125 | ||
| X80333.1 | - | 3 - 4 | 189 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab18
Genomic Map