Assay Details
| Gene Symbol: Rps6ka2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011299.4 | NP_035429.1 | 7 - 8 | 834 | 60 | |
| GenBank mRNA | AJ131021.1 | - | 7 - 8 | 662 | 60 | |
| AK019881.1 | - | 7 - 8 | 739 | 60 | ||
| AK132079.1 | - | 6 - 7 | 502 | 60 | ||
| BC038251.1 | - | 1 - 2 | 61 | 60 | ||
| BC043064.1 | - | 7 - 8 | 784 | 5706153 | 60 | |
| BC051079.1 | - | 7 - 8 | 744 | 6491001 | 60 | |
| BC055331.1 | - | 7 - 8 | 834 | 6406879 | 60 | |
| BC056946.1 | - | 7 - 8 | 777 | 6822831 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka2
Genomic Map