Assay Details
| Gene Symbol: Rps6ka2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011299.4 | NP_035429.1 | 15 - 16 | 1657 | 67 | |
| GenBank mRNA | AJ131021.1 | - | 15 - 16 | 1485 | 67 | |
| AK019881.1 | - | 15 - 16 | 1562 | 67 | ||
| BC038251.1 | - | 9 - 10 | 884 | 67 | ||
| BC043064.1 | - | 15 - 16 | 1607 | 5706153 | 67 | |
| BC055331.1 | - | 15 - 16 | 1657 | 6406879 | 67 | |
| BC056946.1 | - | 15 - 16 | 1600 | 6822831 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka2
Genomic Map