Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700005694.mm.1 |
NM_001205214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005694.mm.1 |
NM_001205214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005694.mm.1 |
NM_011306.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005694.mm.1 |
NM_011306.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005694.mm.1 |
TC1700000611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005693.mm.1 |
NM_001205214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005693.mm.1 |
NM_001205214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005693.mm.1 |
NM_011306.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005693.mm.1 |
NM_011306.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005693.mm.1 |
TC1700000611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005702.mm.1 |
NM_001205214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005702.mm.1 |
NM_001205214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005702.mm.1 |
NM_001205215.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005702.mm.1 |
NM_001205215.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005702.mm.1 |
NM_001205216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005702.mm.1 |
NM_001205216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005702.mm.1 |
NM_011306.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005702.mm.1 |
NM_011306.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005702.mm.1 |
TC1700000611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005696.mm.1 |
NM_001205214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005696.mm.1 |
NM_001205214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005696.mm.1 |
NM_011306.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005696.mm.1 |
NM_011306.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005696.mm.1 |
TC1700000611.mm.1 |
MTA TC |