Assay Details
| Gene Symbol: Rxrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205214.1 | NP_001192143.1 | 3 - 4 | 786 | 67 | |
| NM_001205215.1 | NP_001192144.1 | 3 - 4 | 509 | 67 | ||
| NM_001205216.1 | NP_001192145.1 | 3 - 4 | 509 | 67 | ||
| NM_011306.4 | NP_035436.1 | 3 - 4 | 786 | 67 | ||
| GenBank mRNA | AK140371.1 | - | 3 - 4 | 525 | 67 | |
| AK159671.1 | - | 3 - 4 | 787 | 67 | ||
| BC019432.1 | - | 3 - 4 | 441 | 5132301 | 67 | |
| BC049773.1 | - | 3 - 4 | 746 | 6581828 | 67 | |
| KJ650219.1 | - | 3 - 4 | 600 | 67 | ||
| KJ663717.1 | - | 3 - 4 | 600 | 67 | ||
| KJ663718.1 | - | 4 - 5 | 486 | 67 | ||
| KJ663719.1 | - | 4 - 5 | 486 | 67 | ||
| KJ663720.1 | - | 3 - 4 | 417 | 67 | ||
| KJ663721.1 | - | 4 - 5 | 471 | 67 | ||
| M26804.1 | - | 2 - 3 | 378 | 67 | ||
| M84818.1 | - | 3 - 4 | 459 | 67 | ||
| X66224.1 | - | 2 - 3 | 378 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rxrb
Genomic Map