Assay Details
| Gene Symbol: Ccl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011338.2 | NP_035468.1 | 1 - 2 | 250 | 93 | |
| GenBank mRNA | AF128195.1 | - | 1 - 2 | 97 | 93 | |
| AF128196.1 | - | 1 - 2 | 97 | 93 | ||
| AF128197.1 | - | 1 - 2 | 97 | 93 | ||
| AF128198.1 | - | 1 - 2 | 97 | 93 | ||
| AF128199.1 | - | 1 - 2 | 97 | 93 | ||
| AF128200.1 | - | 1 - 2 | 97 | 93 | ||
| AF128201.1 | - | 1 - 2 | 97 | 93 | ||
| AF128202.1 | - | 1 - 2 | 97 | 93 | ||
| AF128203.1 | - | 1 - 2 | 97 | 93 | ||
| AF128204.1 | - | 1 - 2 | 97 | 93 | ||
| AK151131.1 | - | 1 - 2 | 118 | 93 | ||
| AK151649.1 | - | 1 - 2 | 118 | 93 | ||
| AK154511.1 | - | 1 - 2 | 118 | 93 | ||
| AK154657.1 | - | 1 - 2 | 118 | 93 | ||
| AK155032.1 | - | 1 - 2 | 118 | 93 | ||
| AK155036.1 | - | 1 - 2 | 118 | 93 | ||
| BC139318.1 | - | 1 - 2 | 102 | 8862340 | 93 | |
| BC145962.1 | - | 1 - 2 | 102 | 40131265 | 93 | |
| CB950873.1 | - | 1 - 2 | 253 | 93 | ||
| U15209.1 | - | 1 - 2 | 231 | 93 | ||
| U19482.1 | - | 1 - 2 | 130 | 93 | ||
| U49513.1 | - | 1 - 2 | 106 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl9
Genomic Map