Assay Details
| Gene Symbol: Cxcl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011339.2 | NP_035469.1 | 1 - 2 | 94 | 66 | |
| GenBank mRNA | AF082859.1 | - | 1 - 2 | 57 | 66 | |
| AK142955.1 | - | 1 - 2 | 96 | 66 | ||
| BC061138.1 | - | 1 - 2 | 87 | 30298837 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cxcl15
Genomic Map