Assay Details
| Gene Symbol: Serpinf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011340.3 | NP_035470.3 | 3 - 4 | 412 | 64 | |
| GenBank mRNA | AF017057.1 | - | 3 - 4 | 310 | 64 | |
| AF036164.1 | - | 3 - 4 | 310 | 64 | ||
| AK012411.1 | - | 3 - 4 | 342 | 64 | ||
| BC019852.1 | - | 3 - 4 | 322 | 5123884 | 64 | |
| D50460.1 | - | 3 - 4 | 381 | 64 | ||
| D87975.1 | - | 3 - 4 | 309 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpinf1
Genomic Map