Assay Details
| Gene Symbol: Sema3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011349.3 | NP_035479.2 | 16 - 17 | 1924 | 63 | |
| GenBank mRNA | AF080090.1 | - | 16 - 17 | 1830 | 63 | |
| AF080091.1 | - | 15 - 16 | 1723 | 63 | ||
| AK053632.1 | - | 16 - 17 | 1925 | 63 | ||
| AK140340.1 | - | 16 - 17 | 2405 | 63 | ||
| BC010976.1 | - | 16 - 17 | 1845 | 4219060 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sema3f
Genomic Map