Assay Details
| Gene Symbol: Sema4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB021291.1 | - | 9 - 10 | 1246 | 123 | |
| AF038652.1 | - | 8 - 9 | 1005 | 123 | ||
| AK035993.1 | - | 9 - 10 | 1278 | 123 | ||
| AK047067.1 | - | 9 - 10 | 1262 | 123 | ||
| AK143196.1 | - | 9 - 10 | 1290 | 123 | ||
| AK158292.1 | - | 9 - 10 | 1212 | 123 | ||
| BC057048.1 | - | 8 - 9 | 1154 | 123 | ||
| BC090625.1 | - | 9 - 10 | 1248 | 123 | ||
| BC094567.1 | - | 9 - 10 | 1273 | 6834138 | 123 | |
| BC139232.1 | - | 9 - 10 | 1220 | 8862254 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sema4f
Genomic Map