Assay Details
| Gene Symbol: Sim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011376.3 | NP_035506.2 | 5 - 6 | 784 | 81 | |
| GenBank mRNA | AB221630.1 | - | 5 - 6 | 538 | 81 | |
| D79209.1 | - | 5 - 6 | 577 | 81 | ||
| U40575.1 | - | 5 - 6 | 78 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sim1
Genomic Map