Assay Details
| Gene Symbol: Slc1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077514.3 | NP_001070982.1 | 2 - 3 | 763 | 84 | |
| NM_001077515.2 | NP_001070983.1 | 2 - 3 | 190 | 84 | ||
| NM_011393.2 | NP_035523.1 | 2 - 3 | 190 | 84 | ||
| GenBank mRNA | AB007810.1 | - | 2 - 3 | 820 | 84 | |
| AB007811.1 | - | 2 - 3 | 179 | 84 | ||
| AB007812.1 | - | 2 - 3 | 190 | 84 | ||
| AK134609.1 | - | 2 - 3 | 764 | 84 | ||
| AK140095.1 | - | 2 - 3 | 758 | 84 | ||
| BC138255.1 | - | 2 - 3 | 190 | 8861274 | 84 | |
| BC138256.1 | - | 2 - 3 | 190 | 8861275 | 84 | |
| D43796.1 | - | 2 - 3 | 343 | 84 | ||
| KC825358.1 | - | 1 - 2 | 152 | 84 | ||
| U11763.1 | - | 2 - 3 | 348 | 84 | ||
| U75372.1 | - | 2 - 3 | 776 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a2
Genomic Map