Assay Details
| Gene Symbol: Slc1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077514.3 | NP_001070982.1 | 3 - 4 | 907 | 70 | |
| NM_001077515.2 | NP_001070983.1 | 3 - 4 | 334 | 70 | ||
| NM_011393.2 | NP_035523.1 | 3 - 4 | 334 | 70 | ||
| GenBank mRNA | AB007810.1 | - | 3 - 4 | 964 | 70 | |
| AB007811.1 | - | 3 - 4 | 323 | 70 | ||
| AB007812.1 | - | 3 - 4 | 334 | 70 | ||
| AK134609.1 | - | 3 - 4 | 908 | 70 | ||
| AK140095.1 | - | 3 - 4 | 902 | 70 | ||
| BC138255.1 | - | 3 - 4 | 334 | 8861274 | 70 | |
| BC138256.1 | - | 3 - 4 | 334 | 8861275 | 70 | |
| D43796.1 | - | 3 - 4 | 487 | 70 | ||
| KC825358.1 | - | 2 - 3 | 296 | 70 | ||
| U11763.1 | - | 3 - 4 | 492 | 70 | ||
| U75372.1 | - | 3 - 4 | 920 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a2
Genomic Map