Assay Details
| Gene Symbol: Slc1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077514.3 | NP_001070982.1 | 9 - 10 | 2023 | 78 | |
| NM_001077515.2 | NP_001070983.1 | 9 - 10 | 1450 | 78 | ||
| NM_011393.2 | NP_035523.1 | 9 - 10 | 1450 | 78 | ||
| GenBank mRNA | AB007810.1 | - | 9 - 10 | 2080 | 78 | |
| AB007811.1 | - | 9 - 10 | 1439 | 78 | ||
| AB007812.1 | - | 9 - 10 | 1450 | 78 | ||
| AK134609.1 | - | 9 - 10 | 2024 | 78 | ||
| BC138255.1 | - | 9 - 10 | 1450 | 8861274 | 78 | |
| BC138256.1 | - | 9 - 10 | 1450 | 8861275 | 78 | |
| D43796.1 | - | 9 - 10 | 1603 | 78 | ||
| KC825358.1 | - | 8 - 9 | 1412 | 78 | ||
| U11763.1 | - | 9 - 10 | 1608 | 78 | ||
| U24699.1 | - | 8 - 9 | 1421 | 78 | ||
| U75372.1 | - | 9 - 10 | 2036 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc1a2
Genomic Map