Assay Details
| Gene Symbol: Slc2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011401.4 | NP_035531.3 | 4 - 5 | 864 | 61 | |
| GenBank mRNA | AK028041.1 | - | 4 - 5 | 638 | 61 | |
| AK045328.1 | - | 4 - 5 | 640 | 61 | ||
| AK147258.1 | - | 3 - 4 | 3176 | 61 | ||
| AK155997.1 | - | 4 - 5 | 623 | 61 | ||
| AK164286.1 | - | 4 - 5 | 636 | 61 | ||
| BC034122.1 | - | 4 - 5 | 622 | 4527667 | 61 | |
| BC050095.1 | - | 4 - 5 | 618 | 61 | ||
| BC058811.1 | - | 4 - 5 | 622 | 6490844 | 61 | |
| CT010354.1 | - | 3 - 4 | 509 | 61 | ||
| M75135.1 | - | 4 - 5 | 588 | 61 | ||
| X61093.1 | - | 4 - 5 | 588 | 61 | ||
| X69698.1 | - | 53 | 61 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Slc2a3
Genomic Map