Assay Details
| Gene Symbol: Slc2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011401.4 | NP_035531.3 | 6 - 7 | 1216 | 77 | |
| GenBank mRNA | AK045328.1 | - | 6 - 7 | 992 | 77 | |
| AK147258.1 | - | 5 - 6 | 3528 | 77 | ||
| AK155997.1 | - | 6 - 7 | 975 | 77 | ||
| AK164286.1 | - | 6 - 7 | 988 | 77 | ||
| BC034122.1 | - | 6 - 7 | 974 | 4527667 | 77 | |
| BC050095.1 | - | 6 - 7 | 970 | 77 | ||
| BC058811.1 | - | 6 - 7 | 974 | 6490844 | 77 | |
| CT010354.1 | - | 5 - 6 | 861 | 77 | ||
| M75135.1 | - | 6 - 7 | 940 | 77 | ||
| X61093.1 | - | 6 - 7 | 940 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc2a3
Genomic Map