Assay Details
| Gene Symbol: Slc4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011403.2 | NP_035533.1 | 11 - 12 | 1636 | 90 | |
| GenBank mRNA | AK137542.1 | - | 11 - 12 | 1532 | 90 | |
| AK157181.1 | - | 11 - 12 | 1605 | 90 | ||
| AK157833.1 | - | 11 - 12 | 1494 | 90 | ||
| AK157861.1 | - | 11 - 12 | 1506 | 90 | ||
| AK158143.1 | - | 11 - 12 | 1637 | 90 | ||
| AK158184.1 | - | 11 - 12 | 1602 | 90 | ||
| AK158243.1 | - | 11 - 12 | 1506 | 90 | ||
| AK158453.1 | - | 11 - 12 | 1481 | 90 | ||
| AK162616.1 | - | 11 - 12 | 1481 | 90 | ||
| AK165195.1 | - | 11 - 12 | 1493 | 90 | ||
| AK165267.1 | - | 11 - 12 | 1500 | 90 | ||
| AK171662.1 | - | 1532 | 90 | |||
| AK171808.1 | - | 11 - 12 | 1470 | 90 | ||
| AK171845.1 | - | 1532 | 90 | |||
| AK172103.1 | - | 11 - 12 | 1493 | 90 | ||
| AK172316.1 | - | 11 - 12 | 1481 | 90 | ||
| AK172395.1 | - | 11 - 12 | 1532 | 90 | ||
| AK172456.1 | - | 11 - 12 | 1516 | 90 | ||
| AK172569.1 | - | 11 - 12 | 1481 | 90 | ||
| AY296129.1 | - | 11 - 12 | 1469 | 90 | ||
| BC052419.1 | - | 11 - 12 | 1476 | 5702599 | 90 | |
| BC053429.1 | - | 11 - 12 | 1467 | 6506270 | 90 | |
| M29379.1 | - | 11 - 12 | 1463 | 90 | ||
| X02677.1 | - | 11 - 12 | 1469 | 90 | ||
| X03917.1 | - | 9 - 10 | 1313 | 90 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK165267.1 transcripts.
Set Membership: |
|

Slc4a1
Genomic Map