Assay Details
| Gene Symbol: Slc4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011403.2 | NP_035533.1 | 15 - 16 | 2238 | 73 | |
| GenBank mRNA | AK137542.1 | - | 15 - 16 | 2134 | 73 | |
| AK157181.1 | - | 15 - 16 | 2207 | 73 | ||
| AK157833.1 | - | 15 - 16 | 2096 | 73 | ||
| AK158143.1 | - | 15 - 16 | 2239 | 73 | ||
| AK158184.1 | - | 15 - 16 | 2204 | 73 | ||
| AK158243.1 | - | 15 - 16 | 2108 | 73 | ||
| AK158453.1 | - | 15 - 16 | 2083 | 73 | ||
| AK162616.1 | - | 15 - 16 | 2083 | 73 | ||
| AK165195.1 | - | 15 - 16 | 2095 | 73 | ||
| AK165267.1 | - | 15 - 16 | 2102 | 73 | ||
| AK171662.1 | - | 2134 | 73 | |||
| AK171808.1 | - | 15 - 16 | 2072 | 73 | ||
| AK171845.1 | - | 2134 | 73 | |||
| AK172103.1 | - | 15 - 16 | 2095 | 73 | ||
| AK172316.1 | - | 15 - 16 | 2083 | 73 | ||
| AK172395.1 | - | 15 - 16 | 2134 | 73 | ||
| AK172456.1 | - | 15 - 16 | 2118 | 73 | ||
| AK172569.1 | - | 15 - 16 | 2083 | 73 | ||
| AY296129.1 | - | 15 - 16 | 2071 | 73 | ||
| BC052419.1 | - | 15 - 16 | 2078 | 5702599 | 73 | |
| BC053429.1 | - | 15 - 16 | 6506270 | 73 | ||
| M29379.1 | - | 15 - 16 | 2065 | 73 | ||
| X02677.1 | - | 15 - 16 | 2071 | 73 | ||
| X03917.1 | - | 13 - 14 | 1915 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a1
Genomic Map