Assay Details
| Gene Symbol: Slc8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112798.2 | NP_001106269.1 | 6 - 7 | 2235 | 73 | |
| NM_001286684.1 | NP_001273613.1 | 8 - 9 | 2271 | 73 | ||
| NM_011406.3 | NP_035536.2 | 8 - 9 | 2271 | 73 | ||
| NR_104580.1 | 6 - 7 | 2270 | 73 | |||
| GenBank mRNA | AF004666.1 | - | 8 - 9 | 2266 | 73 | |
| AK048160.1 | - | 1733 | 73 | |||
| BC079673.1 | - | 5 - 6 | 2126 | 30532156 | 73 | |
| DQ388998.1 | - | 5 - 6 | 2160 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc8a1
Genomic Map