Assay Details
| Gene Symbol: Spa17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011449.2 | NP_035579.1 | 3 - 4 | 416 | 107 | |
| GenBank mRNA | AK005571.1 | - | 3 - 4 | 285 | 107 | |
| BC059727.1 | - | 3 - 4 | 286 | 6775177 | 107 | |
| BU936068.1 | - | 3 - 4 | 421 | 107 | ||
| CK137444.1 | - | 3 - 4 | 277 | 107 | ||
| Z46299.1 | - | 3 - 4 | 316 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spa17
Genomic Map