Assay Details
| Gene Symbol: Stxbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011504.1 | NP_035634.1 | 15 - 16 | 1391 | 61 | |
| GenBank mRNA | AK138609.1 | - | 15 - 16 | 1423 | 61 | |
| AK138769.1 | - | 3 - 4 | 674 | 61 | ||
| AK151466.1 | - | 15 - 16 | 1386 | 61 | ||
| AK152261.1 | - | 15 - 16 | 1380 | 61 | ||
| AK155133.1 | - | 15 - 16 | 1389 | 61 | ||
| AK169799.1 | - | 14 - 15 | 1311 | 61 | ||
| BC062901.1 | - | 15 - 16 | 1400 | 6810254 | 61 | |
| BC094055.1 | - | 15 - 16 | 1399 | 61 | ||
| D30798.1 | - | 15 - 16 | 1391 | 61 | ||
| U19521.1 | - | 15 - 16 | 1516 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stxbp3
Genomic Map