Assay Details
| Gene Symbol: Syk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198977.1 | NP_001185906.1 | 12 - 13 | 1914 | 62 | |
| NM_011518.2 | NP_035648.2 | 12 - 13 | 1918 | 62 | ||
| GenBank mRNA | AK049207.1 | - | 12 - 13 | 1943 | 62 | |
| AK136363.1 | - | 12 - 13 | 1917 | 62 | ||
| AK143564.1 | - | 12 - 13 | 1947 | 62 | ||
| BC065121.1 | - | 12 - 13 | 1850 | 30533300 | 62 | |
| U25685.1 | - | 11 - 12 | 1702 | 62 | ||
| U36776.1 | - | 12 - 13 | 1921 | 62 | ||
| Z49877.1 | - | 12 - 13 | 2176 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syk
Genomic Map