Assay Details
| Gene Symbol: Tgfb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011577.1 | NP_035707.1 | 4 - 5 | 1578 | 79 | |
| GenBank mRNA | AJ009862.1 | - | 4 - 5 | 1578 | 79 | |
| AK144163.1 | - | 4 - 5 | 1580 | 79 | ||
| BC013738.1 | - | 4 - 5 | 1025 | 3586216 | 79 | |
| M13177.1 | - | 4 - 5 | 1063 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfb1
Genomic Map