Assay Details
| Gene Symbol: Timm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011592.2 | NP_035722.2 | 3 - 4 | 325 | 65 | |
| GenBank mRNA | AK153866.1 | - | 3 - 4 | 329 | 65 | |
| BC023454.1 | - | 3 - 4 | 301 | 65 | ||
| BC043479.1 | - | 3 - 4 | 305 | 65 | ||
| BC110677.1 | - | 3 - 4 | 323 | 4979986 | 65 | |
| BC117523.1 | - | 3 - 4 | 315 | 40105563 | 65 | |
| BC117524.1 | - | 3 - 4 | 315 | 40105561 | 65 | |
| CA493459.1 | - | 3 - 4 | 363 | 65 | ||
| U69898.1 | - | 3 - 4 | 315 | 65 | ||
More Information
Set Membership: |
|

Timm44
Genomic Map